Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® 96 Sample Purification System

VWR® 96 Sample Purification System

VWR® 96 Tips combs CS50, 96 Tip Combs are designed to be used with the VWR 96 Sample Processing System.
Catalog # 77321-882
Supplier:   VWR International
VWR® 96 Sample Purification System
Catalog # 77321-882
Supplier:  VWR International
VWR® 96 Sample Purification System
VWR® 96 Sample Purification System

VWR® 96 Tips combs CS50, 96 Tip Combs are designed to be used with the VWR 96 Sample Processing System.

Price
$437.83
Log in for availability
Case of 50
Product Details & Documents

VWR® 96 Sample Purification System is a high throughput, automated system that improves the efficiency and reproducibility of magnetic bead-based purification protocols.

  • Compatible with all popular magnetic bead kits
  • On-board programming with a touch screen control panel
  • Process up to 96 samples in deep-well plates
  • 8 position carousel with 2 heating positions
  • Sample volume 50 to 1000 µl

Capable of processing up to 96 samples at one time, the VWR® 96 Sample purification system is easy to set up and program. Mixing, magnetic bead transfer, washing, and elution steps are performed automatically. The purification process can be completed within 10 to 60 minutes depending on the type of reagent kit used and complexity of the program.


Programming
The VWR® 96 Sample purification system can be programmed by using the integrated touchscreen or the included mouse. No external PC is required. After a protocol has been created, the instrument will calculate and display the estimated time for each step and the total processing time.


Typical step settings include:
Tip comb loading and unloading: Automate the tip comb loading and unloading by designating the respective plate positions in the script.
Mixing: Parameters for mixing time, speed, and amplitude can be programmed for each step of the protocol.
Temperature control: Heating of the sample mixture during the lysis and elution steps significantly improves the purity and yield of the target molecule. The VWR® 96 Sample purification system is equipped with 2 heating blocks on the plate carousel that can be precisely set to temperatures from ambient +5 to 120 °C. Deep well plates fit into the heating blocks for efficient heat transfer. A cooling fan inside the processing chamber maintains the ambient operating temperature to ensure consistent results.
Bead collection and release: The complete collection of magnetic beads from sample mixtures is crucial for maximizing the yield of purified analytes. The system’s software offers full control of magnetic parameters to ensure cleaner extractions. General settings allow control of magnet speed, position, repeat collection cycles, etc., to guarantee consistent yields.


Protocol Optimization
The VWR® 96 Sample purification system has been validated with several magnetic bead kits from leading manufacturers and a number of optimized protocols for specific applications are available. To request scripts for specific kits, or for assistance in optimization, please contact VWR® Technical Support Team at [email protected].


Chamber UV Sterilization
An ultraviolet sterilization lamp (UVC) inside the processing chamber provides effective elimination of most bacterial, viral, and stray genomic DNA contaminants. This additional safeguard reduces the possibility of unwanted DNA/RNA contamination of critical samples that could be amplified downstream in a PCR detection assay. UV sterilization time can be adjusted based on specific program requirements to ensure a sterile environment prior to purification.


Communication Ports
The VWR® 96 Sample purification system can store up to 100 program files which can be transferred between instruments, or to a computer for storage or sharing. Simply insert the included flash drive to export the selected files. Programming control can be done via the touchscreen, or by using the included mouse. A PC can also be connected for remote programming.


VWR® Consumables
VWR® offers high-quality deep well plates and magnetic tip combs designed to fit the VWR® 96 Sample purification system.

Certifications: CE Mark

Specifications
  • Description:
    VWR® 96 Tips combs CS50
  • Details:
    96 Tip Combs are designed to be used with the VWR 96 Sample Processing System.
Complementary Products

Product Information

  • Catalog #
    Description
    Color
    Well Volume
    Sterility
    Availability
    Price
  • 43001-0504
    96-well plate, square wells, V-bottom
    Transparent
    2.0 ml
    Non sterile
    Log in for availability
    Case of 60 - $970.66

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.