CARTRIDGE CLARIFLOW E MINI
Catalog # 77320-700
Supplier: PARKER HANNIFIN TE
CARTRIDGE CLARIFLOW E MINI
Catalog # 77320-700
Supplier: PARKER HANNIFIN TE
Supplier Number: 25-10110-002-41-E
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Direct-zol™ RNA Kits, Zymo Research
Supplier: Zymo Research
Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers
Supplier: Ricca Chemical
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Preserved Starfish
Supplier: Avantor
DIBMA Screening Kit, HEPES
Supplier: CUBE BIOTECH
VWR® General Purpose Dissecting Scissors
Supplier: VWR International
E 5 LIQ MACH DISHWSH UN 4L
Supplier: Ostrem Chemical
Buffer Solution
Supplier: Ward's Science
Ponceau S solution 0.1% in 1% acetic acid protein stain
Supplier: Azure Biosystems
Ascorbic Acid
Supplier: Ward's Science
E.Z.N.A.® Plasmid Mini Kit I, Omega Bio-tek®
Supplier: Omega Bio-Tek
Accessories for Gas Regulators
Supplier: VWR International
Acetic acid glacial 99.5-100.5% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC