FILTER CAPSULE PROFLOW II 0.1UM PTFE
Catalog # 77320-716
Supplier: PARKER HANNIFIN TE
FILTER CAPSULE PROFLOW II 0.1UM PTFE
Catalog # 77320-716
Supplier: PARKER HANNIFIN TE
Supplier Number: 34-CDEPP-001-N
Frequently Bought Together
D-(+)-Sucrose, Ultrapure DNase-, RNase-free
Supplier: VWR
Starswabs II® Specimen Collection and Transport Systems
Supplier: Starplex
VWR® EZ-Vision® Dye-as-Loading Buffer, 6X
Supplier: VWR
Calcium chloride, anhydrous ≥96% ACS
Supplier: VWR
Leakbuster™ Specimen Containers
Supplier: Starplex
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Nunc-Immuno™ MicroWell™ 96-Well Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
Difco™ Agar Soldifying Agent, BD Diagnostics
Supplier: BD
Specialty Cell Culture Media
Supplier: Lonza
Orion™ Sure-Flow® pH Electrode 9172BNWP, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
Supplier: VWR International
Biebrich scarlet
Supplier: Thermo Scientific Chemicals
Cerium(IV) sulfate solution 0.100 N (N/10) in sulphuric acid 2N
Supplier: Ricca Chemical
Acetaldehyde 99%
Supplier: Thermo Scientific Chemicals
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo