10-Level Conductivity Indicators
10-level conductivity indicator
Catalog # 470352-982
Supplier: GSC International
Supplier #: CNDTEST
Jump to:
Product Details & Documents
A very simple and visual demonstration of the level of conductivity of almost any solution.
- Simple to use
- Battery included and can be replaced as needed
- LED Indicator light are a quick visual confirmation of the level of conductivity
- Two pack sizes available
The 10-Level conductivity indicator can be used for quick comparisons of conductivity in solutions including, but not limited to, tap water, vinegar, baking soda solution, or varying concentrations of salt water. This device is very simple to use, just submerge both brass prongs in a solution hold the button on the side, and wait for the bar of LED's to display your results. Instruction are included.
Caution: Prop 65 Warning Label is listed on the package for lead and BPA.
Specifications
- Description:10-level conductivity indicator