$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
BIN WIRE MESH STK/HNG 7-1/4X4-1/4X3 CS20
77037-228
:
QUANTUM STORAGE SYSTEMS
BIN WIRE MESH STK/HNG 7-1/4X4-1/4X3 CS20
77037-228
:
QUANTUM STORAGE SYSTEMS
:
QMB520CCS
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Universal Pipette Tips
: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH