Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BIN STK/HNG PP BLACK 10-7/8X5-1/2X5 CS12
Catalog # 77037-478
Supplier:
QUANTUM STORAGE SYSTEMS
Expand image
BIN STK/HNG PP BLACK 10-7/8X5-1/2X5 CS12
Catalog # 77037-478
Supplier:
QUANTUM STORAGE SYSTEMS
Supplier Number:
QUS230BKCS
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH