$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
BOX DIVID GRID GRAY 22.5X17.5X12 CS3
77037-186
:
QUANTUM STORAGE SYSTEMS
BOX DIVID GRID GRAY 22.5X17.5X12 CS3
77037-186
:
QUANTUM STORAGE SYSTEMS
:
DG93120GYCS
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Universal Pipette Tips
: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH