Bee-Write™ ESD Cleanroom Papers
ESD cleanroom paper, Green, 8.5×11", 250 sheets/ream
Supplier: High-Tech Conversions
Supplier #: BW-CRP-8511-GREEN
Jump to:
Product Details & Documents
Bee-Write™ ESD Cleanroom Paper is cleanroom packaged (vacuum packed, double-bagged) and processed in a class 100 ISO 5 controlled environment.
- Low Chloride and other ionic contamination; ultra-low ionic contamination
- Formulated without Magnesium Silicate
- Excellent IPA resistance
- Compatible to most inks in writing without smearing
- Ultra-low particle and fiber generation
- Excellent heat resistance material for laser printing and photocopiers
- Compatible with laser and inkjet printers
- Latex-free
- ROHS compliant
Because Bee-Write™ Cleanroom Paper is ESD safe, it is also suitable for use in a variety of electronics assembly cleanrooms. It is compatible with most pens as well as laser and inkjet printers.
Specifications
- Color:Green
- Description:ESD cleanroom paper
- Dimensions:8.5×11"
- Packaging:250 sheets/ream