Microscope Cover Glasses, Rectangular, No. 1
18×18 mm, 0.13 - 0.16 mm, Super white glass
76970-088
Supplier #: SCG218118
Four E's premium cover glasses with available selection between super white glass or borosilicate glass 3.3
- Precision-cut for uniform sized pieces
- Ultra-clear glass for optimal visibility
- Premium quality material for durability
- Autoclavable
No. 1 pertains to 0.13 to 0.16 mm thickness. The super white glass offering allows for superior light transmittance for enhanced visibility.
- L×W:18×18 mm
- Material:Super white glass
- Thickness:0.13 - 0.16 mm