Orders LinkContactUsLinkComponent
Control Slides, Normal Human Tissue, Kidney
Control slides, normal human tissue, kidney
  76948-238
 :   Mopec
 :  SC950-100
Control Slides, Normal Human Tissue, Kidney
  76948-238
 :  Mopec
Control Slides, Normal Human Tissue, Kidney
Control Slides, Normal Human Tissue, Kidney
Control slides, normal human tissue, kidney
$2,054.79
Case of 100
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using formalin fixed paraffin embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
 : Case of 25 contains 24 unstained slides and 1 stained slide. Case of 100 contains 98 unstained slides and 2 stained slides.
  • Description:
    Control slides, normal human tissue, kidney
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)