Orders LinkContactUsLinkComponent
PIP® Dynamic Bump Caps
Bump cap, HDPE, Hi-vis orange
  76944-676
 :   PIP
 :  280-HP940-31
PIP® Dynamic Bump Caps
  76944-676
 :  PIP
PIP® Dynamic Bump Caps
PIP® Dynamic Bump Caps
Bump cap, HDPE, Hi-vis orange
$6.95
Each Case of 20
Bump Caps feature a four-point plastic suspension and pin-lock adjustable back.
  • Lightweight ventilated HDPE shell
  • Four-point HDPE pin-lock suspension
  • With foam sweatband
Popular in food processing, manufacturing, maintenance, and service industries, the bump cap is designed as a safeguard against accidental bumping or scraping of the head.
Compatible face shield bracket 251-EPB940 and Visor 251-FA815. Compatible with chin strap 280-HP241C. Does not meet and is not tested to any safety standards.
  • Color:
    Hi-vis orange
  • Description:
    Bump cap
  • Material:
    HDPE