Order Entry
Canada
Orders LinkContactUsLinkComponent
 

1321N1 Stable Cells, Human, Multispan

MULTISCREEN™ Human PAR1 HeLa HTS Stable Cell Line, Cryopreserved, Single donor, 2 Vials
Catalog # 76918-900
Supplier:   MULTISPAN INC
 :  C1207-7
1321N1 Stable Cells, Human, Multispan
Catalog # 76918-900
Supplier:  MULTISPAN INC
CAS Number:  
1321N1 Stable Cells, Human, Multispan
1321N1 Stable Cells, Human, Multispan

MULTISCREEN™ Human PAR1 HeLa HTS Stable Cell Line, Cryopreserved, Single donor, 2 Vials

CAS Number:  
Price
$58,965.20
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Specifications

Specifications

  • Cell Format:
    Cryopreserved
  • Single Donor/Pooled:
    Single donor
  • Description:
    MULTISCREEN™ Human PAR1 HeLa HTS Stable Cell Line
  • Size:
    2 Vials
  • Cat. No.:
    76918-900
  • Storage conditions:
    LN₂ (Below –135 ℃)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.