Orders LinkContactUsLinkComponent
RSA-Pro X™ Micro-Sample Management Systems and Inserts | Avantor
RSA-Pro X™ Micro-Sample Management Systems and Inserts
Micro-Sample Management System. Complete set, includes 96 individual, 1 ml glass Inserts with RSA-Pro X™ surfaces in a rack and base plus a matching silicone rubber, Pre-Slit plug style, sealing mat
Catalog # 76881-744
Fournisseur #:  95296-1ML-3PP
RSA-Pro X™ Micro-Sample Management Systems and Inserts
Catalog # 76881-744
RSA-Pro X™ Micro-Sample Management Systems and Inserts
Micro-Sample Management System. Complete set, includes 96 individual, 1 ml glass Inserts with RSA-Pro X™ surfaces in a rack and base plus a matching silicone rubber, Pre-Slit plug style, sealing mat
Price
205,64 $
Log in for availability
Each
La quantité minimale de commande est de 1
Product Details & Documents
Specifications
  • Description:
    Micro-Sample Management System. Complete set, includes 96 individual, 1 ml glass Inserts with RSA-Pro X™ surfaces in a rack and base plus a matching silicone rubber, Pre-Slit plug style, sealing mat
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.