FLOW CONTROLLER (FID GASSES FOR GC-FID)
Catalog # 76836-988
Supplier: LUCIDITY SYSTEMS LLC
FLOW CONTROLLER (FID GASSES FOR GC-FID)
Catalog # 76836-988
Supplier: LUCIDITY SYSTEMS LLC
Supplier Number: P100176
Frequently Bought Together
Leakbuster™ Specimen Containers
Supplier: Starplex
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
AccuStart™ II PCR SuperMix, QuantaBio
Supplier: Quantabio
SYRINGE WITH NEEDLE 5ML
Supplier: AMX INDUSTRIAL TRADE MX
Glycerin
Supplier: Ward's Science
D-(+)-Sucrose
Supplier: G-Biosciences
Acetone, low water content ≥99.3%, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics
TrueBlot® IP/Western Blot, Rockland Immunochemicals
Supplier: Rockland Immunochemical
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Difco™ Nutrient Agar, BD
Supplier: BD
Potassium Phosphate Monobasic
Supplier: Ward's Science
Sodium peroxide, granular for analysis, Supelco®
Supplier: MilliporeSigma
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
Test Kits, COD, TNT plus®
Supplier: Hach