LEAK SENSOR FOR LC-UV
Catalog # 76836-948
Supplier: LUCIDITY SYSTEMS LLC
Frequently Bought Together
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Gastight® 1700 Series Syringes, Hamilton
Supplier: Hamilton
Hydrochloric acid (2.87 - <lt/> 7.8 mol/l; 2.87 - <lt/> 7.8 N) 6.0 N analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
2-Propanol ≥99.8% (by GC), HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Camco® Erado-Sol® Biological and Chemical Stain Remover, Cambridge
Supplier: Cambridge Diagnostic Products
Phage Display Competent Cells, Biosearch Technologies
Supplier: Lucigen
SingleQuots® EGM®-2 Endothelial Kit
Supplier: LONZA PHARMA - BIOSCIENCE CA
DpnI, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs (NEB)
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton
Cardboard Freezer Boxes and Dividers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Sodium Dodecyl Sulfate
Supplier: Ward's Science
Single-Use Needles, BD Medical
Supplier: BD