ACCU-SCOPE® EXC-120 Upright Microscopes
Product Details & Documents
The EXC-120 Series is engineered for classroom, medical and veterinary applications.
Binocular, monocular or trinocular viewing head
Digital microscope options
Head is rotatable 360°
Available with Darkfield contrasting
DIN 4×, 10×, SP40×R and 100×R oil objectives
Rackless mechanical stage
Delivers sharp, high contrast objectives, ergonomic operation, and a rugged, die-cast aluminum alloy frame, the EXC-120 series will deliver excellent performance through years of heavy use. ACCU-SCOPE® offers the EXC-120 series with the options of brightfield observation, darkfield observation, phase contrast, simple polarization, in a trinocular, binocular, monocular or dual view observation viewing head, and the option of adding a camera for your digital microscope options.
Specifications
- Magnification:4×, 10×, 40×R, 100×R
- Description:Four-objective digital trinocular HD
- Camera:Excelis™ HD LITE
- Cat. No.:470351-282
- Observation method:Brightfield
- Electrical:Corded