$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
LID USE W/24 OR 96-WELL PLATES CS100
76808-458
:
THOMSON INSTRUMENT CO
LID USE W/24 OR 96-WELL PLATES CS100
76808-458
:
THOMSON INSTRUMENT CO
:
981945
About Thomson Instrument
Thomson Instrument Company Collection and Filter Plates
Thomson Instrument Product Catalog
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma