Optimum Growth® Sampling and Multiport Flasks
Product Details & Documents
Thomson’s Optimum Growth® Sampling and multiport flasks are engineered for contamination-free sampling, aseptic feeding, and sterile media transfer, making them ideal for seed train development, upstream bioprocessing, and closed-system cell culture maintenance. These flasks eliminate the need for rocker bioreactors, transfer containers, and tube fusing setups while maintaining sterility and shaker compatibility throughout the workflow.
Available in multiple sizes
- One-way sampling valve allows safe and aseptic sampling while the flask remains in the shaker or on the benchtop
- Streamlines workflows and minimizes time, contamination risk, and handling complexity
- Closed system designs helps maintain sterility throughout use, reducing contamination risk and supporting GMP compliance
Together, these flasks deliver a powerful combination of ease-of-use, contamination control, and operational flexibility, making them a leading choice for modern upstream bioprocessing workflows.
Thomson’s Optimum Growth® Sampling Flasks feature integrated one-way valves that enable aseptic sampling directly in the shaker or on the benchtop, helping reduce contamination risk and improve efficiency, especially in high-throughput environments. These flasks are available in multiple sizes with integrated one-way sampling valves, allowing aseptic sampling without removing the cap. Their design streamlines workflows and minimizes time, contamination risk, and handling complexity. The 0.2 µm PTFE vented caps support sterile gas exchange and maintain culture integrity.
Thomson’s Optimum Growth® Multiport flasks offer integrated ports that enable closed-system feeding, sampling, and media transfer, supporting sterile workflows without the need to open the flask, even during extended culture periods. These flasks are available in multiple sizes with a one-way sampling valve that allows safe and aseptic sampling while the flask remains in the shaker or on the benchtop. A two-way Luer lock port supports sterile feeding, inoculation, and direct media transfer to larger vessels with no tube fusing required, as media can be added or withdrawn through integrated ports. Their design eliminates the need for cell bags up to 50 L, reducing cost and complexity. Compatible with multi-day batch feeding in shaker environments, multiport flasks include sterile, pre-attached tubing for plug-and-play integration into cleanroom workflows.
Specifications
- Cap type:Screw cap
- Sterility:Sterile
- Capacity:500 ml
- Color:Translucent
- Description:Multiport Optimum Growth™ flask with threaded top, dual port vent cap, Male Luer lock connections, 381 µl sample tubing, C-Flex® 16 tubing (¹/₄"Ext.Ø.×¹/₈"Int.Ø)
- Tubing length:24" (609.6 mm)
Accessories
Product Information
- Catalog #DimensionsAvailabilityPrice
- 8-position carrier for 125 ml Optimum Growth® flasks8-position carrier for 125 ml Optimum Growth® flasks76808-36010.75×5"Log in for availabilityEach - 1 210,00 $
Product Information
- Catalog #DimensionsAvailabilityPrice
- 8-position carrier for 250 ml Optimum Growth® flasks8-position carrier for 250 ml Optimum Growth® flasks76808-36213.4×6"Log in for availabilityEach - 1 210,00 $
Product Information
- Catalog #ØAvailabilityPrice
- Stand with ring for inverting 1.6 and 2.8 L Optimum Growth® flasksStand with ring for inverting 1.6 and 2.8 L Optimum Growth® flasks76808-5145" ringLog in for availabilityEach - 340,00 $
Product Information
- Catalog #ØAvailabilityPrice
- Stand with ring for inverting 5 L Optimum Growth® flasksStand with ring for inverting 5 L Optimum Growth® flasks76808-5107" ringLog in for availabilityEach - 340,00 $
Product Information
- Catalog #ØAvailabilityPrice
- Ring only for inverting 1.6 and 2.8 L Optimum Growth® flasksRing only for inverting 1.6 and 2.8 L Optimum Growth® flasks76808-5165" ringLog in for availabilityEach - 170,00 $
Product Information
- Catalog #ØAvailabilityPrice
- Ring only for inverting 5 L Optimum Growth® flasksRing only for inverting 5 L Optimum Growth® flasks76808-5127" ringLog in for availabilityEach - 170,00 $
Product Information
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® vent caps for 500 ml flasksOptimum Growth® vent caps for 500 ml flasks76808-548SterileLog in for availabilityCase of 25 - 387,00 $
Product Information
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® 1.6 L flask and bioreactor exhaust capsOptimum Growth® 1.6 L flask and bioreactor exhaust caps76808-330SterileLog in for availabilityCase of 8 - 2 210,00 $
Product Information
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® vent caps for 1.6 L, 2.8 L, and 5 L flasksOptimum Growth® vent caps for 1.6 L, 2.8 L, and 5 L flasks76808-554SterileLog in for availabilityCase of 24 - 603,00 $
Product Information
- Catalog #MaterialSterilityAvailabilityPrice
- Rapid Clear® cap 3000
Product Information
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® vent caps for 125 ml flasksOptimum Growth® vent caps for 125 ml flasks76808-544SterileLog in for availabilityCase of 50 - 641,00 $
Product Information
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® vent caps for 250 ml flasksOptimum Growth® vent caps for 250 ml flasks76808-546SterileLog in for availabilityCase of 50 - 641,00 $
Product Information
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® low evaporation caps for 1.6, 2.8, and 5 L flasksOptimum Growth® low evaporation caps for 1.6, 2.8, and 5 L flasks76808-354SterileLog in for availabilityCase of 24 - 415,00 $