WHEATON® Serum Vials
Serum vial, 5 ml, clear EP, tray, 5 ml, 21.75×40 mm, Glass, Clear, No
: WUBB5MLC-T-EP
Serum vials converted from Type I borosilicate glass tubing.
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
- Crimp cap type
Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.
- Cap included:No
- Capacity:5 ml
- Color:Clear
- Description:Serum vial, 5 ml, clear EP, tray
- Material:Glass
- Diameter ×H:21.75×40 mm



