WHEATON® Serum Vials
Serum vial, 3 ml, clear EP, tray, 3 ml, 16.75×37.7 mm, Glass, Clear, No
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: WUBB3MLC-T-EP
Jump to:
Product Details & Documents
Serum vials converted from Type I borosilicate glass tubing.
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
- Crimp cap type
Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.
Documents
Specifications
- Cap included:No
- Capacity:3 ml
- Color:Clear
- Description:Serum vial, 3 ml, clear EP, tray
- Material:Glass
- Diameter ×H:16.75×37.7 mm



