Order Entry
Canada
Orders LinkContactUsLinkComponent
WHEATON® Serum Vials

WHEATON® Serum Vials

Serum vial,10 ml, clear EP, tray, 10 ml, 23.75×50 mm, Glass, Clear, No
Catalog # 76716-350
Supplier #:  WUBB10MLC-T-EP
WHEATON® Serum Vials
Catalog # 76716-350
WHEATON® Serum Vials
WHEATON® Serum Vials

Serum vial,10 ml, clear EP, tray, 10 ml, 23.75×50 mm, Glass, Clear, No

Price
$266.63
New Customer Promotional Price
Log in for availability
Case of 150
Product Details & Documents

Serum vials converted from Type I borosilicate glass tubing.

  • Shrink-wrapped modules reduce particulate contamination
  • Special design adds strength for freeze-drying applications
  • Crimp cap type

Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.

Specifications
  • Cap included:
    No
  • Capacity:
    10 ml
  • Color:
    Clear
  • Description:
    Serum vial,10 ml, clear EP, tray
  • Material:
    Glass
  • Diameter ×H:
    23.75×50 mm