Order Entry
Canada
Orders LinkContactUsLinkComponent
WHEATON® Serum Vials

WHEATON® Serum Vials

Serum vial, 10 ml, amber EP, tray, 10 ml, 23.75×50 mm, Glass, Amber, No
Catalog # 76716-342
Supplier #:  WUBB10MLA-T-EP
WHEATON® Serum Vials
Catalog # 76716-342
WHEATON® Serum Vials
WHEATON® Serum Vials

Serum vial, 10 ml, amber EP, tray, 10 ml, 23.75×50 mm, Glass, Amber, No

Price
$323.05
New Customer Promotional Price
Log in for availability
Case of 150
Product Details & Documents

Serum vials converted from Type I borosilicate glass tubing.

  • Shrink-wrapped modules reduce particulate contamination
  • Special design adds strength for freeze-drying applications
  • Crimp cap type

Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.

Specifications
  • Cap included:
    No
  • Capacity:
    10 ml
  • Color:
    Amber
  • Description:
    Serum vial, 10 ml, amber EP, tray
  • Material:
    Glass
  • Diameter ×H:
    23.75×50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.