FILTERS PP MURUS 1UM GI PLPP301BBPSL
Catalog # 76683-330
Supplier: PARKER HANNIFIN TE
FILTERS PP MURUS 1UM GI PLPP301BBPSL
Catalog # 76683-330
Supplier: PARKER HANNIFIN TE
Supplier Number: PLPP3-01BB-PSL
Frequently Bought Together
Leakbuster™ Specimen Containers
Supplier: Starplex
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Self-Seal Sterilization Pouches
Supplier: VWR International
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
Sulfuric acid standardised solution 1.0 N in aqueous solution
Supplier: Thermo Scientific Chemicals
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
VACUUBRAND® ME 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Supplier: Brandtech
Saline, 0.85%
Supplier: Hardy Diagnostics
Ammonia (Nitrogen) Test Strips, Hach
Supplier: Hach
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
1-Cetylpyridinium chloride monohydrate 98%
Supplier: Thermo Scientific Chemicals
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Immunoreagents
Potassium hydroxide 0.100 N (N/10) in isopropanol (IPA)
Supplier: Ricca Chemical
Giemsa Stain
Supplier: Ward's Science
Lactobacilli MRS Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA