FILTERS PP MURUS 0.6UM PLPP296BBPNL
Catalog # 76683-430
Supplier: PARKER HANNIFIN TE
FILTERS PP MURUS 0.6UM PLPP296BBPNL
Catalog # 76683-430
Supplier: PARKER HANNIFIN TE
Supplier Number: PLPP2-96BB-PNL
Frequently Bought Together
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
METHYLPARABEN 500G
Supplier: VWR
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals
Hydrogen peroxide solution 30%, AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
VACUUBRAND® ME 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Supplier: Brandtech
Saline, 0.85%
Supplier: Hardy Diagnostics
Nickel(II) chloride hexahydrate 98%
Supplier: Thermo Scientific Chemicals
Mounting Bracket For Qlicksmart® BladeFLASK Blade Remover, Mortech
Supplier: Mortech Manufacturing
Glycerine (natural) ≥99.0% USP
Supplier: Spectrum Chemical Mfg. Corp.
BBL™ Brain Heart Infusion, BD Biosciences
Supplier: BD
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC