FILTERS PP MURUS 3UM GI PLPP103BBPSL
76683-350
Frequently Bought Together
Water, deionized VWR®
: VWR
n-Hexane ≥95%, OmniSolv®, Supelco®
: MilliporeSigma
Methanol ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
: MilliporeSigma
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
: Thermo Fisher Scientific
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
: VWR International
Gastight® 1700 Series Syringes, Hamilton
: Hamilton
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
Corning® Mouse Laminin
: Corning
LabKlenz™ 100 Alkaline Detergents
: Steris
Sol-M® Hypodermic Needles
: SOL-MILLENNIUM CANADA, INC. CA
VWR® Reagent alcohol
: VWR International