FILTERS PP MURUS 1UM GI PLPP101BBPSL
Catalog # 76683-394
Supplier: PARKER HANNIFIN TE
FILTERS PP MURUS 1UM GI PLPP101BBPSL
Catalog # 76683-394
Supplier: PARKER HANNIFIN TE
Supplier Number: PLPP1-01BB-PSL
Frequently Bought Together
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Methanol ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
VWR® Specimen Containers, PP with PE Caps
Supplier: VWR International
RNA Clean & Concentrator™ Kits, Zymo Research
Supplier: Zymo Research
Single-Use Needles, BD Medical
Supplier: BD
Fendall® Saline Eyewash Bottles, Honeywell Safety
Supplier: Honeywell Safety Products
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
GelRed® & GelGreen® Nucleic Acid Gel Stains, Biotium
Supplier: Biotium
Puritan PurFlock Ultra® Flocked Swabs, Puritan Medical Products
Supplier: Puritan Medical Products
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Ligation Sequencing Kits
Supplier: Oxford Nanopore Technologies
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning
Quantofix Chloride Test Strips, CTL Scientific Supply
Supplier: Macherey-Nagel
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Supplier: Starplex
Flip-Top Containers, Capitol Vial®
Supplier: Thermo Fisher Scientific