FILTERS PP MURUS 0.6UM GI PLPP396BBPSL
Catalog # 76683-342
Supplier: PARKER HANNIFIN TE
FILTERS PP MURUS 0.6UM GI PLPP396BBPSL
Catalog # 76683-342
Supplier: PARKER HANNIFIN TE
Supplier Number: PLPP3-96BB-PSL
Frequently Bought Together
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
Water, PCR grade
Supplier: VWR International
Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
Supplier: Cardinal Health
Fendall® Saline Eyewash Bottles, Honeywell Safety
Supplier: Honeywell Safety Products
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
Vesphene® III st Disinfectant Cleaners
Supplier: Steris
Hydrochloric acid 3.00 N NF, USP
Supplier: Ricca Chemical
HistoGel™ Specimen Processing Gel, Epredia
Supplier: Thermo Scientific
SELECT GR FBS CR ORIG 500ML
Supplier: VWR
Reagent Sets for Colorimeters, Hach
Supplier: Hach
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Ward's® Live<i> E. coli </i>cultures for Coliphage Use
Supplier: Avantor