FILTERS PP MURUS 10UM GI PLPP210AAPSL
Catalog # 76683-346
Supplier: PARKER HANNIFIN TE
FILTERS PP MURUS 10UM GI PLPP210AAPSL
Catalog # 76683-346
Supplier: PARKER HANNIFIN TE
Supplier Number: PLPP2-10AA-PSL
Frequently Bought Together
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
COD Digestion vials, VWR Chemicals BDH®
Supplier: VWR International
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
9-Fluorenylmethyl chloroformate (Fmoc chloride) 98+%
Supplier: Thermo Scientific Chemicals
Puritan® MALDI-TOF Pointed Applicator Stick, Puritan Medical Products
Supplier: Puritan Medical Products
Phusion High-Fidelity DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
SWAB CLEANER KIT PK50
Supplier: Advanced Instruments
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Bovine Serum Albumin (BSA)
Supplier: Rockland Immunochemical
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton