Elevated Zero Mazes
The elevated zero maze is an elevated, ring-shaped runway with the same amount of area devoted to adjacent open and closed quadrants, similar to the elevated plus maze to study anxiety behaviors.
- Acrylic construction
- Odor-free
- Matted finish for non-reflective surfaces
- Easy to clean
- Easy assembly and disssembly with a removable base
The elevated zero maze is very similar to the elevated plus maze, but with the center region of the elevated plus maze removed. It has been pharmacologically validated with various anxiolytic drugs.
The design is created with an elevated circular platform with two walled, enclosed portions as well as two open portions, allowing for clear interpretations of murine behavior. Because mice tend to avoid open-spaced areas, the animals will preferentially spend more time in the enclosed walled area.
- Ø:50 cm
- Width:5 cm
- Description:Elevated zero maze; mouse
- Height:20 cm (wall), 61 cm (stand)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)