Bite Test Chambers
Product Details & Documents
The Zebrafish bite test evaluates cerebral lateralization, which promotes the use of a particular eye.
- Clear acrylic construction
Zebrafish bite test evaluates cerebral lateralization which promotes the use of a particular eye, according to the visual stimulus that is being fixated. Additionally, the bite test apparatus is employed to assess and correlate the biting habituation in Zebrafish to the preferential eye use, on viewing a novel object.
The apparatus can be conveniently attached to the inside of a home tank. The bite test apparatus stays connected to the main part of the tank with the help of a small opening that allows the fish to enter or leave the box during trials expediently.
Specifications
- Width:12 cm
- Description:Bite test with home chamber, zebrafish
- Height:3.5 cm
- Length:15 cm
- Material:Acrylic
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



