Dispenser Dropping Bottles
Dispenser dropping bottle, 30 ml, LDPE
Catalog # 470349-766
Supplier: United Scientific Supplies
Supplier #: UNDDB030
Jump to:
Product Details & Documents
Precision dispensing of solutions.
- Dispenses one drop at a time
- Spout lid is connected to the cap
- LDPE with leakproof PP cap
- BPA-free
This LDPE dispensing bottle can be easily operated with one hand. The bottle features a polypropylene screw cap with a tethered spout lid that snaps securely in place.
Specifications
- Capacity:30 ml
- Description:Dispenser dropping bottle
- Material:LDPE