Orders LinkContactUsLinkComponent
Dispenser Dropping Bottles
Dispenser Dropping Bottles
Dispenser dropping bottle, 15 ml, LDPE
Catalog # 470349-746
Supplier #:  UNDDB015
Dispenser Dropping Bottles
Catalog # 470349-746
Dispenser Dropping Bottles
Dispenser Dropping Bottles
Dispenser dropping bottle, 15 ml, LDPE
Price
$82.50
Log in for availability
Pack of 72
Product Details & Documents
Precision dispensing of solutions.
  • Dispenses one drop at a time
  • Spout lid is connected to the cap
  • LDPE with leakproof PP cap
  • BPA-free
This LDPE dispensing bottle can be easily operated with one hand. The bottle features a polypropylene screw cap with a tethered spout lid that snaps securely in place.
Specifications
  • Capacity:
    15 ml
  • Description:
    Dispenser dropping bottle
  • Material:
    LDPE