Orders LinkContactUsLinkComponent
Slide Storage Boxes, Hinged, Plastic
Slide storage box, hinged, Plastic, 25 slides
  470349-718
 :  62211
Slide Storage Boxes, Hinged, Plastic
  470349-718
Slide Storage Boxes, Hinged, Plastic
Slide Storage Boxes, Hinged, Plastic
Slide storage box, hinged, Plastic, 25 slides
$7.25
Each
Store slides securely and safely.
  • Engraved numbers
  • Secure closing clasp
  • Key card for easy reference
Sturdy two-piece molded slide boxes hold standard 3×1" microscope slides. The hinged top has a reference card for labeling the corresponding slide description. The numbers on the opposite side are engraved for easy reference. These boxes are made from high impact polystyrene.
  • Capacity:
    25 slides
  • Description:
    Slide storage box, hinged
  • Material:
    Plastic