Order Entry
Canada
Orders LinkContactUsLinkComponent
DNA Markers

DNA Markers

DNA marker, 35 and 1500 bp, 3.2 ml, For use with the Fragment analyzer systems and the DNF-910 kit
Catalog # CA76645-396
Supplier #:  FS-SMK910-0003
DNA Markers
Catalog # CA76645-396
CAS Number:  
DNA Markers

DNA marker, 35 and 1500 bp, 3.2 ml, For use with the Fragment analyzer systems and the DNF-910 kit

CAS Number:  
Price
$309.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Description:
    DNA marker
  • Size:
    35 and 1500 bp
  • Volume:
    3.2 ml
  • Details:
    For use with the Fragment analyzer systems and the DNF-910 kit

Specifications