Order Entry
Canada
Orders LinkContactUsLinkComponent
DNA/RNA Ladders

DNA/RNA Ladders

DNA ladder, 100 - 1500 bp, 3200 µl, For use with the Fragment analyzer systems and the DNF-910, DNF-935 kits or the ZAG DNA analyzer system and the ZAG-135 kits
Catalog # CA76645-070
Supplier #:  FS-SLR910-0003
DNA/RNA Ladders
Catalog # CA76645-070
CAS Number:  
DNA/RNA Ladders
DNA/RNA Ladders

DNA ladder, 100 - 1500 bp, 3200 µl, For use with the Fragment analyzer systems and the DNF-910, DNF-935 kits or the ZAG DNA analyzer system and the ZAG-135 kits

CAS Number:  
Price
$309.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Description:
    DNA ladder
  • Size:
    100 - 1500 bp
  • Volume:
    3200 µl
  • Details:
    For use with the Fragment analyzer systems and the DNF-910, DNF-935 kits or the ZAG DNA analyzer system and the ZAG-135 kits

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.