RNA ScreenTape Analysis
Product Details & Documents
Reliable, efficient, and objective measurements of quality and quantity of RNA samples.
- Utilizes RINe - a user independent algorithm for accurate and objective assessment of eukaryotic or prokaryotic total RNA integrity
- Provides fast results with data delivered in approximately 1 minute per sample
- Minimal sample consumption is required as only 1 µl of sample is needed for accurate data
- RNA sample detection down to 5 ng/µl is possible
- Objective quality control of precious RNA samples is enabled within RNAseq, microarray analysis, and quantitative RT-PCR workflows
- Ready-to-use consumables provide excellent reproducibility
- No cross contamination and sample carryover by using single lanes and individual loading tips
- Partially used ScreenTape devices can be used for completion at a later point in time
RNA ScreenTape analysis provides efficient and reliable electrophoretic separation of total RNA down to 5 ng/µl. Compatible with eukaryotic and prokaryotic samples, the assay provides accurate information about sample integrity and quantity. Objective evaluation of RNA degradation is delivered with an RNA Integrity Number (RINe) that can be used for sample quality control in NGS workflows.
The assay is compatible with all Agilent TapeStation instruments.
Note: Some products within this assay contain SYBR™ Gold, which is licensed from Life Technologies Corporation for use in research and development only. SYBR™ is a registered trademark of Life Technologies Corporation.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Specifications
- Description:RNA ScreenTape ladder
- Details:For the analysis of total RNA down to a sensitivity of 5 ng/µl, ladder only, to order with 5067-5576 and 5067-5577