Order Entry
Canada
Orders LinkContactUsLinkComponent
 

DNA/RNA Ladders

Extended genomic DNA ladder, 75 - 48500 bp, 50 µl, 250 ng/µl, For use with the fragment analyzer systems and the DNF-467 kit
  CA76645-320
 :  DNF-367-U050
DNA/RNA Ladders
  CA76645-320
 :  
DNA/RNA Ladders
DNA/RNA Ladders

Extended genomic DNA ladder, 75 - 48500 bp, 50 µl, 250 ng/µl, For use with the fragment analyzer systems and the DNF-467 kit

 :  
 
$616.00
   
Each
 
 
 

 

  • Concentration:
    250 ng/µl
  • Description:
    Extended genomic DNA ladder
  • Size:
    75 - 48500 bp
  • Volume:
    50 µl
  • Details:
    For use with the fragment analyzer systems and the DNF-467 kit

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.