Order Entry
Canada
Orders LinkContactUsLinkComponent
 

DNA/RNA Ladders

HS extended large frag DNA ladder, 75 - 48500 bp, 125 µl, 1.5 ng/µl, For use with the fragment analyzer systems and the DNF-464 kit
  CA76645-000
 :  DNF-365-U125
DNA/RNA Ladders
  CA76645-000
 :  
DNA/RNA Ladders
DNA/RNA Ladders

HS extended large frag DNA ladder, 75 - 48500 bp, 125 µl, 1.5 ng/µl, For use with the fragment analyzer systems and the DNF-464 kit

 :  
 
$309.00
   
Each
 
 
 

 

  • Concentration:
    1.5 ng/µl
  • Description:
    HS extended large frag DNA ladder
  • Size:
    75 - 48500 bp
  • Volume:
    125 µl
  • Details:
    For use with the fragment analyzer systems and the DNF-464 kit