Order Entry
Canada
Orders LinkContactUsLinkComponent
DNA/RNA Ladders

DNA/RNA Ladders

HS extended genomic DNA ladder, 75 - 48500 bp, 125 µl, 10 ng/µl, For use with the fragment analyzer systems and the DNF-468 kit
Catalog # CA76645-368
Supplier #:  DNF-364-U125
DNA/RNA Ladders
Catalog # CA76645-368
CAS Number:  
DNA/RNA Ladders
DNA/RNA Ladders

HS extended genomic DNA ladder, 75 - 48500 bp, 125 µl, 10 ng/µl, For use with the fragment analyzer systems and the DNF-468 kit

CAS Number:  
Price
$309.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Specifications

Specifications

  • Concentration:
    10 ng/µl
  • Description:
    HS extended genomic DNA ladder
  • Size:
    75 - 48500 bp
  • Volume:
    125 µl
  • Details:
    For use with the fragment analyzer systems and the DNF-468 kit

Specifications