Orders LinkContactUsLinkComponent
Capillary Arrays
Capillary Arrays
FA 12x-capillary array ultra-short, For use with the fragment analyzer 5200 system
Catalog # CA76645-078
Supplier #:  M5310AA-001
Capillary Arrays
Catalog # CA76645-078
Capillary Arrays
Capillary Arrays
FA 12x-capillary array ultra-short, For use with the fragment analyzer 5200 system
Price
Inquire For Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Capillary arrays for fragment analyzer and ZAG systems.
  • Array cartridges are robust and have a long lifespan, keeping your instrument on-task from one project to the next
  • Short arrays provide robust separation for most samples
  • Speed up separation times and increase overall QC throughput with ultra-short arrays
  • Enhance separation resolution and focus nucleic acid analysis with long arrays
The fragment analyzer systems offer a selection of capillary arrays to optimize your QC workflow by separating nucleic acids via parallel capillary electrophoresis. Arrays vary in capillary length, providing distinct benefits to a wide range of workflows.
These arrays address separation speed and separation resolution. 'Short' capillary arrays are the most versatile as they are compatible with all consumable kits. Run times can be decreased with 'Ultra-Short' arrays, and 'Long' arrays enhance separation resolution. Some kits have been designed for a specific array for optimal performance.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
  • Description:
    FA 12x-capillary array ultra-short
  • Details:
    For use with the fragment analyzer 5200 system