Order Entry
Canada
Orders LinkContactUsLinkComponent
DNA Markers

DNA Markers

DNA marker, 75 and 20000 bp, 3.2 ml, For use with the Fragment analyzer systems and the DNF-930 kit or the ZAG DNA analyzer system and the ZAG-130 kit
Catalog # CA76645-034
Supplier #:  FS-SMK930-0003
DNA Markers
Catalog # CA76645-034
CAS Number:  
DNA Markers

DNA marker, 75 and 20000 bp, 3.2 ml, For use with the Fragment analyzer systems and the DNF-930 kit or the ZAG DNA analyzer system and the ZAG-130 kit

CAS Number:  
Price
$309.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Description:
    DNA marker
  • Size:
    75 and 20000 bp
  • Volume:
    3.2 ml
  • Details:
    For use with the Fragment analyzer systems and the DNF-930 kit or the ZAG DNA analyzer system and the ZAG-130 kit

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.