Order Entry
Canada
Orders LinkContactUsLinkComponent
 

DNA/RNA Ladders

HS NGS DNA ladder, 100 - 3000 bp, 100 µl, 1000 pg/µl, For use with the Fragment analyzer systems and the DNF-474 kit
Catalog # CA76645-438
 :  DNF-396-U100
DNA/RNA Ladders
Catalog # CA76645-438
CAS Number:  
DNA/RNA Ladders
DNA/RNA Ladders

HS NGS DNA ladder, 100 - 3000 bp, 100 µl, 1000 pg/µl, For use with the Fragment analyzer systems and the DNF-474 kit

CAS Number:  
Price
$309.00
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1000 pg/µl
  • Description:
    HS NGS DNA ladder
  • Size:
    100 - 3000 bp
  • Volume:
    100 µl
  • Details:
    For use with the Fragment analyzer systems and the DNF-474 kit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.