Order Entry
Canada
Orders LinkContactUsLinkComponent
 

DNA/RNA Ladders

HS RNA ladder, 200 - 6000 nt, 15 µl, 25 ng/µl, For use with the Fragment analyzer systems and the DNF-472 kit
Catalog # CA76645-214
 :  DNF-386-U015
DNA/RNA Ladders
Catalog # CA76645-214
CAS Number:  
DNA/RNA Ladders
DNA/RNA Ladders

HS RNA ladder, 200 - 6000 nt, 15 µl, 25 ng/µl, For use with the Fragment analyzer systems and the DNF-472 kit

CAS Number:  
Price
$309.00
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Concentration:
    25 ng/µl
  • Description:
    HS RNA ladder
  • Size:
    200 - 6000 nt
  • Volume:
    15 µl
  • Details:
    For use with the Fragment analyzer systems and the DNF-472 kit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.