Orders LinkContactUsLinkComponent
Capillary Arrays
FA 48-capillary array short, 33 cm, For use with the fragment analyzer 5300 system
  CA76645-234
 :  A2300-4850-3355
Capillary Arrays
  CA76645-234
Capillary Arrays
Capillary Arrays
FA 48-capillary array short, 33 cm, For use with the fragment analyzer 5300 system
$4,650.00
Each
Capillary arrays for fragment analyzer and ZAG systems.
  • Array cartridges are robust and have a long lifespan, keeping your instrument on-task from one project to the next
  • Short arrays provide robust separation for most samples
  • Speed up separation times and increase overall QC throughput with ultra-short arrays
  • Enhance separation resolution and focus nucleic acid analysis with long arrays
The fragment analyzer systems offer a selection of capillary arrays to optimize your QC workflow by separating nucleic acids via parallel capillary electrophoresis. Arrays vary in capillary length, providing distinct benefits to a wide range of workflows.
These arrays address separation speed and separation resolution. 'Short' capillary arrays are the most versatile as they are compatible with all consumable kits. Run times can be decreased with 'Ultra-Short' arrays, and 'Long' arrays enhance separation resolution. Some kits have been designed for a specific array for optimal performance.
 : For research use only. Not for use in diagnostic procedures.
  • Description:
    FA 48-capillary array short, 33 cm
  • Details:
    For use with the fragment analyzer 5300 system