Order Entry
Canada
Orders LinkContactUsLinkComponent
DNA/RNA Ladders | Avantor

DNA/RNA Ladders

MDK DNA ladder, 100 - 3000 bp, 1200 µl, —, For use with the Fragment analyzer systems and the DNF-910CP kit
Catalog # CA76645-220
Fournisseur #:  FS-SLR480-0001
DNA/RNA Ladders
Catalog # CA76645-220
CAS Number:  
DNA/RNA Ladders
DNA/RNA Ladders

MDK DNA ladder, 100 - 3000 bp, 1200 µl, —, For use with the Fragment analyzer systems and the DNF-910CP kit

CAS Number:  
Price
$309.00
Log in for availability
Each
La quantité minimale de commande est de 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Concentration:
  • Description:
    MDK DNA ladder
  • Size:
    100 - 3000 bp
  • Volume:
    1200 µl
  • Details:
    For use with the Fragment analyzer systems and the DNF-910CP kit

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.