Order Entry
Canada
Orders LinkContactUsLinkComponent
Diluent Markers

Diluent Markers

FP US NGS diluent marker, 1 bp lower marker, 6000 bp upper marker, 25 µl, For use with the Femto pulse system and the FP-1101 kit, 1 vial
Catalog # CA76645-490
Supplier #:  FP-8101-U025
Diluent Markers
Catalog # CA76645-490
CAS Number:  
Diluent Markers

FP US NGS diluent marker, 1 bp lower marker, 6000 bp upper marker, 25 µl, For use with the Femto pulse system and the FP-1101 kit, 1 vial

CAS Number:  
Price
$616.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    1 vial
  • Description:
    FP US NGS diluent marker, 1 bp lower marker, 6000 bp upper marker
  • Volume:
    25 µl
  • Details:
    For use with the Femto pulse system and the FP-1101 kit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.