Order Entry
Canada
Orders LinkContactUsLinkComponent
DNA/RNA Ladders

DNA/RNA Ladders

FP US RNA ladder, 200 - 6000 nt, 35 µl, 2000 pg/µl, For use with the Femto Pulse system and the FP-1201 kit
Catalog # CA76645-262
Supplier #:  FP-7201-U015
DNA/RNA Ladders
Catalog # CA76645-262
CAS Number:  
DNA/RNA Ladders
DNA/RNA Ladders

FP US RNA ladder, 200 - 6000 nt, 35 µl, 2000 pg/µl, For use with the Femto Pulse system and the FP-1201 kit

CAS Number:  
Price
$385.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Concentration:
    2000 pg/µl
  • Description:
    FP US RNA ladder
  • Size:
    200 - 6000 nt
  • Volume:
    35 µl
  • Details:
    For use with the Femto Pulse system and the FP-1201 kit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.