Order Entry
Canada
Orders LinkContactUsLinkComponent
DNA/RNA Ladders

DNA/RNA Ladders

BAC, DNA ladder, 75 - 48500 bp, 35 µl, 3 ng/µl, For use with the Femto Pulse system and the FP-1004 kit
Catalog # CA76645-194
Supplier #:  FP-7004-U035
DNA/RNA Ladders
Catalog # CA76645-194
CAS Number:  
DNA/RNA Ladders
DNA/RNA Ladders

BAC, DNA ladder, 75 - 48500 bp, 35 µl, 3 ng/µl, For use with the Femto Pulse system and the FP-1004 kit

CAS Number:  
Price
$385.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Concentration:
    3 ng/µl
  • Description:
    BAC, DNA ladder
  • Size:
    75 - 48500 bp
  • Volume:
    35 µl
  • Details:
    For use with the Femto Pulse system and the FP-1004 kit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.