Order Entry
Canada
Orders LinkContactUsLinkComponent
DNA Markers

DNA Markers

DNA marker, 1 and 500 bp, 3.2 ml, For use with the fragment analyzer systems and the DNF-905 kit or the ZAG DNA analyzer system and the ZAG-105 kit
Catalog # CA76645-300
Supplier #:  FA-MRK900F-0003
DNA Markers
Catalog # CA76645-300
CAS Number:  
DNA Markers

DNA marker, 1 and 500 bp, 3.2 ml, For use with the fragment analyzer systems and the DNF-905 kit or the ZAG DNA analyzer system and the ZAG-105 kit

CAS Number:  
Price
$309.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Description:
    DNA marker
  • Size:
    1 and 500 bp
  • Volume:
    3.2 ml
  • Details:
    For use with the fragment analyzer systems and the DNF-905 kit or the ZAG DNA analyzer system and the ZAG-105 kit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.