Plasmid DNA Kit
Plasmid DNA analysis kit, 500 samples, Fragment analyzer system kit for the qualitative analysis of supercoiled plasmids from 2000 - 10000 bp, includes reagents necessary for running 500 samples
Catalog # CA76645-276
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Supplier #: DNF-940-K0500
Jump to:
Product Details & Documents
Quality control analysis of supercoiled and linear plasmid DNA for downstream applications.
- Reliable sizing- optimized ladder for the accurate sizing of supercoiled plasmids
- Concentration range - 0.1 to 1 ng/ul
- Fast analysis - QC results in only 30 minutes
- Accurate sizing - sizing range from 2,000 to 10,000 bp for supercoiled plasmids
The plasmid DNA kit is used in the analysis of supercoiled and linear plasmid DNA. The ladder provided with this kit is optimized for the accurate sizing and relative quantification of supercoiled plasmids between 2,000 and 10,000 bp. Linearized plasmids can be evaluated for quality and relative quantification, however only comparative sizing is possible.
: For research use only. Not for use in diagnostic procedures.
Specifications
- Description:Plasmid DNA analysis kit, 500 samples
- Details:Fragment analyzer system kit for the qualitative analysis of supercoiled plasmids from 2000 - 10000 bp, includes reagents necessary for running 500 samples